Learn More
Sodium L-(+)-tartrate dihydrate, 99%
Description
Sodium L-(+)-tartrate dihydrate is used in buffers for molecular biology and cell culture applications. It is also used as an emulsifier and a binding agent in food products such as jellies, margarine, and sausage casingsand also as a food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium L-(+)-tartrate dihydrate is used in buffers for molecular biology and cell culture applications. It is also used as an emulsifier and a binding agent in food products such as jellies, margarine, and sausage casingsand also as a food additive.
Solubility
It is soluble in water (290 mg/ml at 20°C). Insoluble in alcohol, and organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼150°C (in -2H2O) |
| Density | 1.82 |
| Odor | Odorless |
| Quantity | 1000 g |
| Beilstein | 6121732 |
| Merck Index | 14,8684 |
| Solubility Information | It is soluble in water (290mg/ml at 20°C). Insoluble in alcohol,and organic solvents. |
| Formula Weight | 230.08 (194.06 Anhydrous) |
| Percent Purity | 99% |
| Chemical Name or Material | Sodium L-(+)-tartrate dihydrate |
Safety and Handling
| EINECSNumber | 212-773-3 |
| RTECSNumber | WW8150000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.