Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium L-(+)-tartrate dihydrate, 99%

Catalog No. aaa1618730
Click to view available options
Quantity:
250 g
1000 g
5000 g

CAS: 6106-24-7 | C4H8Na2O8 | 230.08 g/mol

Sodium L-(+)-tartrate dihydrate is used in buffers for molecular biology and cell culture applications. It is also used as an emulsifier and a binding agent in food products such as jellies, margarine, and sausage casingsand also as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium L-(+)-tartrate dihydrate is used in buffers for molecular biology and cell culture applications. It is also used as an emulsifier and a binding agent in food products such as jellies, margarine, and sausage casingsand also as a food additive.

Solubility
It is soluble in water (290 mg/ml at 20°C). Insoluble in alcohol, and organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
PubChem CID 131855972
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

Melting Point ∼150°C (in -2H2O)
Density 1.82
Odor Odorless
Quantity 250 g
Beilstein 6121732
Merck Index 14,8684
Solubility Information It is soluble in water (290mg/ml at 20°C). Insoluble in alcohol,and organic solvents.
Formula Weight 230.08 (194.06 Anhydrous)
Percent Purity 99%
Chemical Name or Material Sodium L-(+)-tartrate dihydrate
EINECSNumber 212-773-3
RTECSNumber WW8150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.