Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Lauryl Sulfate, NF, EP, BP, JP, Spectrum™ Chemical

Click to view available options
Quantity:
125 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Sodium Lauryl Sulfate, NF, EP, BP, JP is used as an emulsifying agent, modified-release agent, penetration enhancer, solubilizing agent, tablet and capsule lubricant. All Spectrum Chemical NF, EP, BP, and JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 1 |
Quantity | 500 g |
Packaging | Poly Bottle |
Water | 0.05 |
Grade | Multi-Compendial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction