Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Meta-Silicate, 9-Hydrate, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

13517-24-3
H18Na2O12Si
284.197
PHIQPXBZDGYJOG-UHFFFAOYSA-N
61639

High quality chemicals for laboratory and specialized industrial use

SMILES O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]
IUPAC Name disodium;dioxido(oxo)silane;nonahydrate
Formula Weight 284.2
Packaging Poly Bottle
Quantity 500g
Chemical Name or Material Sodium Meta-Silicate, 9-Hydrate
Grade BAKER ANALYZED™ Reagent