Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Metabisulfite, Anhydrous, Granular, Reagent, ACS, 97%, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Sodium Metabisulfite, Anhydrous, Granular, Reagent, ACS , also known as disodium or metabisulfite, is typically used as an antioxidant, disinfectant or preservative agent. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications
Specifications
CAS Max % | 1 |
Quantity | 6 x 500 g |
Percent Purity | 97% |
Packaging | Poly Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction