Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Metabisulfite, puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog No. 6002367 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
50 kg
6 x 2.5 kg

Catalog No. 60-023-67

Supplier: Honeywell Chemicals 314485KG

Only null left
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. Ph. Eur., dry, 98-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Physical Form Dry
Quantity 5 kg
Linear Formula Na2S2O5
UN Number NONH for all modes of transport
Formula Weight 190.11g/mol
Percent Purity 98 to 100.5%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium Metabisulfite, Reag. Ph. Eur.
Hazard Statement P280-P301 + P312 + P330-P305 + P351 + P338 + P310
Precautionary Statement H302-H318
EINECSNumber 231-673-0
RTECSNumber UX8225000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.