Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Metal
SCI_ED

Catalog No. s25556a Shop All Fisherbrand Products
Click to view available options
Quantity:
10 g
100 g
Packaging:
Lab Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7440-23-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
Synonym natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
PubChem CID 5360545
ChEBI CHEBI:26708
IUPAC Name sodium
SMILES [Na]

Specifications

Color Silver
Atomic Weight 22.99
Density 0.9684 at 20°C
Linear Formula Na
Quantity 100 g
Formula Weight 22.99
Grade Laboratory
Packaging Lab Grade
Chemical Name or Material Sodium Metal
ShelfLife 2 years
DOTInformation DOT Class 4.3, I, : Dangerous When Wet
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.