Sodium metaperiodate, 98%, Alfa Aesar™

Source of Periodic Acid, As An Analytical Reagent, and As An Oxidizing Agent

Manufacturer: Alfa Aesar 1379822

Quantity 100g

 View more versions of this product

Catalog No. AA1379822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-28-5
    Physical Form Powder
    Assay Percent Range 98%
    Molecular Formula INaO4
    Formula Weight 213.89
    MDL Number MFCD00003534
    Synonym Sodium periodate
    Chemical Name or Material Sodium metaperiodate

    Specifications

    Physical Form Powder
    Formula Weight 213.89
    Quantity 100g
    Color Colorless
    DOT Information Hazard Class: 5.1, Packing Group: II
    Density 4.174g/cm3
    EINECS Number 232-197-6
    Odor Odorless
    Assay Percent Range 98%
    Molecular Formula INaO4
    Appearance Colorless powder
    MDL Number MFCD00003534
    CAS 7790-28-5
    Synonym Sodium periodate
    Health Hazard 1 H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Sodium metaperiodate

    H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit