Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium metatungstate hydrate, crystalline

Catalog No. ac304090500
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 652154-12-6 | H2Na6O40W12·xH2O | 2986.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652154-12-6
Molecular Formula H2Na6O40W12·xH2O
MDL Number MFCD00150663
Synonym Sodium polytungstate Hydrate

Specifications

Color White to Yellow
Physical Form Crystalline Powder or Beads
Quantity 50 g
Linear Formula Na6[H2W12O40]·xH2O
Solubility Information Solubility in water: >1000g/L (20°C).
Formula Weight 2986.13
Percent Purity ≥85% (WO3)
Packaging Glass bottle
Chemical Name or Material Sodium metatungstate hydrate
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed. Causes serious eye damage. Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.