Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium metavanadate

Catalog No. 72060500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-060-500G 500 g
72-060-100G 100 g
72-060-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 72-060-500G Supplier Sigma Aldrich Supplier No. 72060500G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium metavanadate (SMV) is a vanadium salt that can be used as a corrosion inhibitor with good inhibition efficiency (98%) at 200 ppm. It can be used in the protection of carbon steel.

Specifications

CAS 13718-26-8
Molecular Formula NaO3V
Linear Formula NaVO3
Quantity 500 g
MDL Number MFCD00011125
Synonym Sodium (meta)vanadate; Sodium trioxovanadate; Sodium vanadate(V); Sodium vanadium oxide; Sodium vanadium trioxide
Molecular Weight (g/mol) 121.93
Percent Purity ≥98.0% (RT)
Biological Activity Respiratory Tract
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.