Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium metavanadate, 96%

Catalog No. p-42127
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle

CAS: 13718-26-8 | NaO3V | 121.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13718-26-8
Molecular Formula NaO3V
Molecular Weight (g/mol) 121.93
MDL Number MFCD00011125
InChI Key CMZUMMUJMWNLFH-UHFFFAOYSA-N
Synonym sodium metavanadate, sodium vanadate v, vanadic acid, monosodium salt, sodium vanadate meta, sodium oxido dioxo vanadium, sodium trioxovanadate 1-, metawanadan sodowy polish, vanadate vo31-, sodium, monosodium trioxovanadate 1-, ccris 6881
PubChem CID 4148882
ChEBI CHEBI:75221
IUPAC Name sodium vanadiumoylolate
SMILES [Na+].[O-][V](=O)=O

Specifications

Melting Point 600°C
Color Beige-Yellow to White or Green
Physical Form Powder
Quantity 250 g
Assay Percent Range 96%
Linear Formula NaVO3
Merck Index 15,8774
Solubility Information Solubility in water: soluble
Formula Weight 121.93
Percent Purity 96%
Packaging Glass bottle
Chemical Name or Material Sodium metavanadate, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Suspected of damaging fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Toxic to aqua
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Call a PO
EINECSNumber 237-272-7
RTECSNumber YW1050000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.