Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium metavanadate, typically 96%, V 38% min

Catalog No. p-4407916
Click to view available options
Quantity:
25 g
250 g
1 kg

CAS: 13718-26-8 | NaO3V | 121.93 g/mol

Sodium metavanadate is used as an analytical reagent and mordant. It is also found to be useful in the photography industry and the pharmaceutical industry. In addition to this, it is used in the making of vanadium alloy and vanadium catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13718-26-8
Molecular Formula NaO3V
Molecular Weight (g/mol) 121.93
MDL Number MFCD00011125
InChI Key CMZUMMUJMWNLFH-UHFFFAOYSA-N
Synonym sodium metavanadate, sodium vanadate v, vanadic acid, monosodium salt, sodium vanadate meta, sodium oxido dioxo vanadium, sodium trioxovanadate 1-, metawanadan sodowy polish, vanadate vo31-, sodium, monosodium trioxovanadate 1-, ccris 6881
PubChem CID 4148882
ChEBI CHEBI:75221
SMILES [Na+].[O-][V](=O)=O

Specifications

Melting Point 630°C
Color White
Physical Form Chunks and/or Powder
Quantity 25 g
Assay Percent Range Typically 96%
Solubility Information Soluble in water.
IUPAC Name sodium;oxido(dioxo)vanadium
Formula Weight 121.93
Odor Odorless
Appearance Off-white to light orange powder
Chemical Name or Material Sodium metavanadate, V 38% min.
Show More Show Less
Hazard Category H301-H319-H332-H361-H372-H500
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P501c
DOTInformation DOT Class: 6.1; Packaging Group: II
EINECSNumber 237-272-7
RTECSNumber YW1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.