Learn More
Sodium metavanadate, typically 96%, V 38% min
Description
Sodium metavanadate is used as an analytical reagent and mordant. It is also found to be useful in the photography industry and the pharmaceutical industry. In addition to this, it is used in the making of vanadium alloy and vanadium catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 630°C |
Color | White |
Physical Form | Chunks and/or Powder |
Quantity | 25 g |
Assay Percent Range | Typically 96% |
Solubility Information | Soluble in water. |
IUPAC Name | sodium;oxido(dioxo)vanadium |
Formula Weight | 121.93 |
Odor | Odorless |
Appearance | Off-white to light orange powder |
Show More |
Safety and Handling
Hazard Category | H301-H319-H332-H361-H372-H500 |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P501c |
DOTInformation | DOT Class: 6.1; Packaging Group: II |
EINECSNumber | 237-272-7 |
RTECSNumber | YW1050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.