Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium methanesulfonate, 99%, Acros Organics™

Click to view available options
Quantity:
5g
100g
500g
2386-57-4
CH4NaO3S
118.09
KKVTYAVXTDIPAP-UHFFFAOYSA-M
sodium methanesulfonate,methanesulfonic acid sodium salt,methanesulfonic acid, sodium salt,sodium methanesulphonate,sodium mesylate,methylsulfonic acid sodium salt,75-75-2 parent,sodium methyl sulfonate,methanesulfonic acid, sodium salt 1:1,sodium methylsulfonate
638112
sodium;methanesulfonate
CS(=O)(=O)[O-].[Na+]
2386-57-4
CH4NaO3S
118.09
KKVTYAVXTDIPAP-UHFFFAOYSA-M
sodium methanesulfonate,methanesulfonic acid sodium salt,methanesulfonic acid, sodium salt,sodium methanesulphonate,sodium mesylate,methylsulfonic acid sodium salt,75-75-2 parent,sodium methyl sulfonate,methanesulfonic acid, sodium salt 1:1,sodium methylsulfonate
638112
sodium;methanesulfonate
CS(=O)(=O)[O-].[Na+]
Linear Formula CH3SO3Na
Solubility Information Solubility in water: soluble.
Formula Weight 118.09
Percent Purity 99%
Free Acid 1% max.
Infrared Spectrum Authentic
Water 1% max.
Quantity 100g
Chemical Name or Material Sodium methanesulfonate

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 219-203-2