Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Methoxide in HPLC Methanol, 0.50 Normal (Sodium Methylate, 0.50 Molar), Reagents

Catalog No. 01333799
Click to view available options
:
Quantity: 4 L

Sodium Methoxide in HPLC Methanol, 0.50 Normal (Sodium Methylate, 0.50 Molar), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 67-56-1, 124-41-4
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Sodium Methylate, Sodium Methanolate, Methoxysodium
IUPAC Name methanol
SMILES CO

Specifications

Boiling Point 65°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 4 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8044
Chemical Name or Material Sodium Methoxide in HPLC Methanol, 0.50 Normal (Sodium Methylate, 0.50 Molar)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.