Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol

Catalog No. AC168600100
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
25 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC168600100 10 L
AC168600250 25 mL
AC168600010 1 L
AC168600025 2.5 L
AC168600251 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC168600100 Supplier Thermo Scientific Chemicals Supplier No. 168600100
Add to Cart

CAS: 124-41-4 | CH3NaO | 54.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-41-4 , 67-56-1
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.02
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Melting Point 1.0°C to 5.0°C
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 93.0°C
Flash Point 33°C
Packaging Plastic drum
Refractive Index 1.3701
Linear Formula CH3ONa
Quantity 10 L
Beilstein 01,28
Fieser 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
Merck Index 15,8775
Specific Gravity 0.97
Solubility Information Solubility in water: reacts. Other solubilities: soluble in methanol
Viscosity 68 mPa.s (20°C)
Formula Weight 54.02
Grade Pure
Physical Form Liquid
Chemical Name or Material Sodium methoxide, 30 wt% Solution in Methanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes damage to organs.
Flammable liquid and vapor.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 204-699-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.