Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Methoxide in Toluene, 0.10 Molar (Sodium Methylate in Toluene, 0.10 Normal), Reagents

Catalog No. 01333800
Click to view available options
:
Quantity: 1 L

Catalog No. 01-333-800

Supplier: Reagents Holdings Llc CS1091001C

Sodium Methoxide in Toluene, 0.10 Molar (Sodium Methylate in Toluene, 0.10 Normal), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 108-88-3, 67-56-1, 124-41-4
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Boiling Point 111°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8666
Chemical Name or Material Sodium Methoxide in Toluene, 0.10 Molar (Sodium Methylate in Toluene, 0.10 Normal)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.