Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Molybdate Dihydrate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Quantity | 5 g |
Linear Formula | Na2MoO4 · 2H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 241.95g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Chemical Name or Material | Sodium Molybdate Dihydrate |
Safety and Handling
EINECSNumber | 231-551-7 |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction