Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium molybdate(VI) dihydrate, 99+%

Catalog No. AC206375000
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle

CAS: 10102-40-6 | MoNa2O4 | 205.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-40-6
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99% min
Linear Formula Na2MoO4·2H2O
Fieser 15,294
Merck Index 15, 8776
Solubility Information Solubility in water: 560g/L (o°C). Other solubilities: 1150g/l (100°C)
Formula Weight 241.95
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 3.28
Density 3.2800g/mL
Chemical Name or Material Sodium molybdate(VI) dihydrate, 99+%
Show More Show Less
RTECSNumber QA5085000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.