Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium molybdenum oxide, anhydrous, Mo 46.2%

Catalog No. p-7048960
Click to view available options
Quantity:
100 g
500 g

CAS: 7631-95-0 | MoNa2O4 | 205.926 g/mol

It is used as an analytical reagent, corrosion inhibitor, and non-oxidizing anodic inhibitor. It reduces the nitrite requirement of fluids inhibited with nitrite-amine and improves the corrosion protection of carboxylate salt fluids. It is utilized in pigments, dyes, anti-freezing agent, cutting oils, metal finishing agent, brightening agent for zinc, additive for fertilizers and micronutrients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-95-0
Molecular Formula MoNa2O4
Molecular Weight (g/mol) 205.926
MDL Number MFCD00003486
InChI Key TVXXNOYZHKPKGW-UHFFFAOYSA-N
Synonym sodium molybdate, disodium molybdate, sodium molybdate vi, natriummolybdat, molybdic acid, disodium salt, natriummolybdat german, sodium molybdate van, sodium molybdate na2moo4, unii-948qaq08i1, ccris 5442
PubChem CID 61424
ChEBI CHEBI:75215
IUPAC Name disodium;dioxido(dioxo)molybdenum
SMILES [O-][Mo](=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 687°C
Color White
Physical Form Powder
Quantity 100 g
Linear Formula Na2MoO4
Merck Index 14,8645
Solubility Information It is soluble in water.
Formula Weight 205.92
Concentration or Composition (by Analyte or Components) Mo 46.2%
Odor Odorless
Refractive Index 1.714
Sensitivity Hygroscopic
Density 3.72 g/mL
Chemical Name or Material Sodium molybdenum oxide, Anhydrous
Show More Show Less
Hazard Category H335-H373
Hazard Statement GHS H Statement
H332
Harmful if inhaled.
Precautionary Statement P260-P271-P304+P340-P312-P314-P501c
EINECSNumber 231-551-7
RTECSNumber QA5085000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.