Learn More
Sodium n-dodecyl sulfate, 99% (dry wt.), water <2%
Description
Denatures and solubilizes proteins for electrophoresis.Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Denatures and solubilizes proteins for electrophoresis.Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.
Solubility
Slightly soluble in ethanol.
Notes
Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | ∽206°C |
Assay Percent Range | 99% (dry wt.) |
Quantity | 100 g |
UN Number | UN2926 |
Beilstein | 3599286 |
Merck Index | 148636 |
Solubility Information | Slightly soluble in ethanol. |
Formula Weight | 288.38 |
Percent Purity | 99% |
Assay | (dry wt.) |
Show More |
Safety and Handling
Hazard Category | Serious eye damage/eye irritation (category 1), Flammable solids (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2), Hazardous to the aquatic environment (chronic category 3), Acute toxicity (category 4) |
Hazard Statement | Causes serious eye damage, Flammable solid, May cause respiratory irritation, May form combustible dust concentrations in air, Causes skin irritation, Harmful to aquatic life with long lasting effects, Harmful if swallowed or if inhaled |
DOTInformation | Hazard Class: 4.1; Packaging Group: III |
EINECSNumber | 205-788-1 |
RTECSNumber | WT1050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.