Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium nitrate, 98+%
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Sodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.
Solubility
Freely soluble in water, ammonia and hydrazine. Soluble in alcohol.Slightly soluble in pyridine.
Notes
Sodium nitrate is hygroscopic in nature.
Sodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.
Solubility
Freely soluble in water, ammonia and hydrazine. Soluble in alcohol.Slightly soluble in pyridine.
Notes
Sodium nitrate is hygroscopic in nature.
Specifications
Specifications
| Melting Point | ∼308°C |
| Quantity | 2500 g |
| UN Number | UN1498 |
| Merck Index | 14,8647 |
| Solubility Information | Freely soluble in water,ammonia and hydrazine. Soluble in alcohol.Slightly soluble in pyridine. |
| IUPAC Name | sodium nitrate |
| Formula Weight | 84.99 |
| Percent Purity | ≥98% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Show More |
Safety and Handling
| Hazard Category | H272-H319-H373 |
| Hazard Statement | GHS H Statement H272-H302 May intensify fire oxidizer. Harmful if swallowed. |
| Precautionary Statement | P210-P220-P221-P260-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: SODIUM NITRATE |
| EINECSNumber | 231-554-3 |
| RTECSNumber | WC5600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction