Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

7631-99-4
NNaO3
84.994
VWDWKYIASSYTQR-UHFFFAOYSA-N
24268
CHEBI:63005
sodium;nitrate
[N+](=O)([O-])[O-].[Na+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 84.99
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.26kg
Quantity 2.5kg
Chemical Name or Material Sodium Nitrate
Grade Baker Analyzed™ ACS Reagent