Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrate Solution, Adjustor for Chloride Specific Ion Electrode, 5.0M (5.0N), LabChem™

Catalog No. LC246652
Click to view available options
Quantity:
500 mL
1 L
4 L

Chemical Identifiers

CAS 7631-99-4, 7732-18-5
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
PubChem CID 24268
ChEBI CHEBI:63005
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Color Colorless
CAS Max % 66.25, 33.75
Linear Formula NaNO3
Concentration 5.0M (5.0N)
Density 1.26g/mL
Solubility Information Soluble in water
Formula Weight 84.99
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.26g/mL
Viscosity 1.4cSt
Decomposition Information Nitrogen oxides; Oxygen
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Sodium Nitrate Solution, Adjustor for Chloride Specific Ion Electrode
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause cancer (oral).
May cause damage to organs (blood, heart, liver) through prolonged or repeated exposure (oral).
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.