Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Nitrite, 10% w/v, Reagents
Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
| Boiling Point | 101°C |
| Color | Colorless |
| pH | 10 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Percent Purity | 10% w/v |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.0539 |
| Chemical Name or Material | Sodium Nitrite, 10% w/v |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction