Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium nitrite, 98%

Catalog No. p-7024287
Click to view available options
Quantity:
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 7632-00-0 | NaNO2 | 69.00 g/mol

Sodium nitrite is used in many industrial applications viz. in meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, an antidote in cyanide poisoning, an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor as well as in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium nitrite is used in many industrial applications viz. in meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, an antidote in cyanide poisoning, an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor as well as in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.

Solubility
Soluble in water, methanol, ethanol, ammonia, nitric acid. Slightly soluble diethyl ether.

Notes
Decomposes above 320°. Hygroscopic. Incompatible with reducing agents, strong oxidizing agents, organics, finely powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NaNO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Melting Point 271°C (decomposition)
Quantity 1000 g
Linear Formula NaNO2
UN Number UN1500
Merck Index 14,8648
Solubility Information Soluble in water,methanol,ethanol,ammonia,nitric acid. Slightly soluble diethyl ether.
Formula Weight 69
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.17
Chemical Name or Material Sodium nitrite
Show More Show Less
Hazard Category H272-H301-H319-H373
Hazard Statement GHS H Statement
H301-H272
Toxic if swallowed.
May intensify fire; oxidizer.
Precautionary Statement P210-P220-P221-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: SODIUM NITRITE
EINECSNumber 231-555-9
RTECSNumber RA1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.