Learn More
Sodium nitrite, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Sodium nitrite is used in many industrial applications viz. in meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, an antidote in cyanide poisoning, an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor and in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 271°C |
Color | White |
Physical Form | Crystals |
Quantity | 25 g |
Assay Percent Range | 99.999% (metals basis) |
Solubility Information | Soluble in water,methanol,ethanol,ammonia,nitric acid. Slightly soluble diethyl ether. |
Formula Weight | 69 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 2.17 g/cm3 at 20°C |
Show More |
Safety and Handling
Hazard Category | H272-H301-H319-H373 |
Precautionary Statement | P210-P220-P221-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c |
DOTInformation | DOT Class: 5.1, Packaging Group: III |
EINECSNumber | 231-555-9 |
RTECSNumber | RA1225000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.