Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrite, puriss. p.a. ACS Reagent, Solstice

Catalog No. 6001512 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-015-12 1 kg
60-015-11 100 g
60-015-13 250 g
60-015-14 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 60-015-12 Supplier Solstice Supplier No. 314431KG
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium;nitrite
SMILES [Na+].[O-]N=O

Specifications

Quantity 1 kg
Linear Formula NaNO2
UN Number 1500
Formula Weight 69g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Ignition Point 914°F
Chemical Name or Material Sodium Nitrite, Reag. Ph. Eur.
Hazard Statement P220-P273-P301 + P310-P305 + P351 + P338
Precautionary Statement H272-H301-H319-H400
EINECSNumber 231-555-9
RTECSNumber RA1225000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.