Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Orthovanadate, 100 mM Solution, PROMO, Thermo Scientific Chemicals

Catalog No. J60191AEPM
Click to view available options
Quantity:
100 mL

Applications

Sodium orthovanadate, is used as an inhibitor of several phosphatases and kinases such as protein tyrosine phosphatases (PTPs), alkaline phosphatase (ALP), and ATPase. It also acts as an inhibitor of adenosine kinase (AK) and phosphofructokinase (PFK). It also suppresses p53-mediated apoptosis by transcription-dependent and -independent pathways.

Solubility

It is soluble in water (50 mg/mL)

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13721-39-6
Molecular Formula Na3O4V
Molecular Weight (g/mol) 183.91 g/mol
MDL Number MFCD00003511
InChI Key IHIXIJGXTJIKRB-UHFFFAOYSA-N
Synonym sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
PubChem CID 61671
ChEBI CHEBI:35607
IUPAC Name trisodium;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 100 mL
Solubility Information It is soluble in water
Concentration or Composition (by Analyte or Components) 100 mM
Chemical Name or Material Sodium orthovanadate
TSCA Yes
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.