Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Oxalate
SCI_ED

Catalog No. s25561 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Reagent Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Color White
Density 2.34g/mL
Assay Percent Range >99.5%
Packaging Reagent Grade
Linear Formula Na2C2O4
Quantity 25 g
Formula Weight 134
Grade Reagent
Chemical Name or Material Sodium Oxalate
ShelfLife 1 year
DOTInformation DOT Class 6.1, III, : Poison
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.