Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium oxalate, ACS, 99.5+%

Catalog No. AA4175930
Click to view available options
Quantity:
50 g
250 g
1 kg

Catalog No. AA4175930

Supplier: Thermo Scientific Chemicals 04175930

Only null left
Add to Cart

CAS: 62-76-0 | C2Na2O4 | 134.00 g/mol

Sodium oxalate is used as a reducing agent, as a primary standard for standardizing potassium permanganate solutions and as a neutralizer for exothermic acidic reactions. It finds application in metal cleaning preparations, metal extraction and separation, blueprint coatings, electroplating baths, special cement manufacturing, plant nutrition as well as in textile and pharmaceutical industries. It plays an important role to remove calcium ions from blood plasma and prevents blood clotting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium;oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Melting Point 250°C to 270°C (decomposition)
Color White
Physical Form Powder
Quantity 250 g
Beilstein 3631622
Merck Index 14,8650
Solubility Information Soluble in water and formic acid; Insoluble in ether and alcohol.
Formula Weight 134
Percent Purity ≥99.5%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Density 2.340 g/mL
Chemical Name or Material Sodium oxalate
Show More Show Less
Hazard Category H302+H312-H320
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
EINECSNumber 200-550-3
RTECSNumber KI1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.