Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium percarbonate, 13-14% active oxygen, Acros Organics

Click to view available options
Quantity:
500g
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
15630-89-4
1·5 H2O2
157.01
MFCD00043204
Sodium carbonate hydrogen peroxide
15630-89-4
1·5 H2O2
157.01
MFCD00043204
Sodium carbonate hydrogen peroxide
Oxidation reagent
Linear Formula 1·5 H2O2
Solubility Information Solubility in water: 150g/L.
Formula Weight 157.01
Physical Form Granular Powder
Percent Purity ≥13% (available oxygen)
Packaging Plastic bottle
Solubility Solubility in water: 150g/L.
Color White
Quantity 2.5kg
Chemical Name or Material Sodium percarbonate

GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 239-707-6