Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium periodate, 99%, for analysis, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
7790-28-5
INaO4
213.89
JQWHASGSAFIOCM-UHFFFAOYSA-M
sodium periodate,sodium metaperiodate,sodium m-periodate,sodiumperiodate,sodium meta periodate,periodic acid, sodium salt,sodium meta-periodate,periodic acid hio4 , sodium salt,periodate sodium,sodium penodate
23667635
CHEBI:75226
sodium;periodate
[O-]I(=O)(=O)=O.[Na+]
7790-28-5
INaO4
213.89
JQWHASGSAFIOCM-UHFFFAOYSA-M
sodium periodate,sodium metaperiodate,sodium m-periodate,sodiumperiodate,sodium meta periodate,periodic acid, sodium salt,sodium meta-periodate,periodic acid hio4 , sodium salt,periodate sodium,sodium penodate
23667635
CHEBI:75226
sodium;periodate
[O-]I(=O)(=O)=O.[Na+]
Oxidation reagent for analysis
Assay Percent Range 98.5% min. (Iodometry)
Linear Formula NaIO4
Fieser 01,809; 02,311; 04,373; 05,507; 06,455; 11,492; 13,145; 15,294
Merck Index 15,8772
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: 389g/L (52°C),soluble in h2so4,hno3,acetic acids,insoluble in most common organic solvents
Formula Weight 213.89
Physical Form Fine Crystals
Percent Purity 99%
Grade Analysis
Chloride 300ppm max
Chlorine (Cl) 300ppm max.
Iodine (I) 10ppm max.
Iron (Fe) 10ppm max.
Lead (Pb) 20ppm max.
Nitrate 2000ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 80g/L (20°C). Other solubilities: 389g/L (52°C), soluble in h2so4, hno3, acetic acids, insoluble in most common organic solvents
Sulfate 200ppm max.
Color White
Melting Point 300.0°C
pH 3.4 to 4.5 (5% soln. at 20°C)
Quantity 500g
Chemical Name or Material Sodium periodate
Show More Show Less

GHS H Statement
May cause fire or explosion; strong oxidizer.
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure if swallowed.
Very toxic to aquatic life.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Danger

EINECSNumber : 232-197-6