Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium perrhenate, 99+% (metals basis), Re 68%

Catalog No. p-4407928
Click to view available options
Quantity:
2 g
10 g
50 g

Catalog No. AA1141204

Supplier: Thermo Scientific Chemicals 01141204

Only null left
Add to Cart

CAS: 13472-33-8 | NaO4Re | 273.193 g/mol

Sodium perrhenate is used as a precursor to other rhenium compounds. Further, it reacts with sodium in ethanol to prepare nonahydridorhenate and with polysulfide solutions to get tetrathiorhenate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-33-8
Molecular Formula NaO4Re
Molecular Weight (g/mol) 273.193
MDL Number MFCD00014231
InChI Key KMISVFTVBOPTAI-UHFFFAOYSA-N
Synonym sodium perrhenate, unii-pe1t8nn47q, sodium tetraoxorhenate 1-, pe1t8nn47q, na.o4re, rhenium vii sodium oxide, rhenate reo41-, sodium, t-4, acmc-1bwm0, perrhenic acid sodium salt, sodium oxido trioxo rhenium
PubChem CID 5107658
IUPAC Name sodium;oxido(trioxo)rhenium
SMILES [O-][Re](=O)(=O)=O.[Na+]

Specifications

Melting Point 300°C
Color Yellow
Physical Form Crystalline
Quantity 2 g
Assay Percent Range 99+% (metals basis)
Solubility Information Soluble in water and ethanol.
Formula Weight 273.19
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline powder
Density 5.39
Chemical Name or Material Sodium perrhenate, Re 68%
Show More Show Less
Hazard Category H272-H315-H319
Precautionary Statement P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 236-742-9
RTECSNumber WD3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.