Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Persulfate, 30% w/v, Reagents

Catalog No. 01333978
Click to view available options
:
Quantity: 4 L

Sodium Persulfate, 30% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 7775-27-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sodium Peroxydisulfate
IUPAC Name water
SMILES O

Specifications

Boiling Point 102°C
Color Colorless
Physical Form Liquid
Quantity 4 L
Percent Purity 30%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1851
Chemical Name or Material Sodium Persulfate, 30% w/v
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.