Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium persulfate, Solstice

Catalog No. 6001542 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-015-42 1 kg
60-015-43 500 g
60-015-44 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-015-42 Supplier Solstice Supplier No. 718901KG

Purum p.a.,≥99.0% (RT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-27-1
Molecular Formula Na2O8S2
Molecular Weight (g/mol) 238.092
MDL Number MFCD00003501
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
PubChem CID 62655
IUPAC Name disodium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 1 kg
Linear Formula Na2S2O8
UN Number UN1505
Formula Weight 238.1g/mol
Percent Purity ≥99% (RT)
Chemical Name or Material Sodium persulfate
Hazard Statement P210-P220-P261-P280-P284-P370 + P378
RTECSNumber SE0525000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.