Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Persulfate (Sodium Peroxydisulfate), Reagents

Catalog No. 01336992
Change view
Click to view available options
Quantity:
500 g
12 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-992 500 g
01-336-995 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-992 Supplier Reagents Holdings Llc Supplier No. C2323100500A5
Add to Cart
Add to Cart

Sodium Persulfate (Sodium Peroxydisulfate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7775-27-1
Molecular Formula Na2O8S2
Molecular Weight (g/mol) 238.09
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
Synonym Sodium Peroxydisulfate
IUPAC Name disodium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.6
Chemical Name or Material Sodium Persulfate (Sodium Peroxydisulfate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.