Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phenoxide, 98%

Catalog No. AAB2500230
Click to view available options
Quantity:
50 g
250 g

CAS: 139-02-6 | C6H5NaO | 116.10 g/mol

Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.

Solubility
Very soluble in water, soluble in alcoholSoluble in water, acetone and alcohol.

Notes
Hygroscopic and air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-02-6
Molecular Formula C6H5NaO
Molecular Weight (g/mol) 116.10
MDL Number MFCD00013134
InChI Key NESLWCLHZZISNB-UHFFFAOYSA-M
Synonym sodium phenoxide, sodium phenolate, sodium phenate, phenolate sodium, sodium benzenolate, sodium carbolate, phenol sodium, phenol sodium salt, phenol, sodium salt, naoph
PubChem CID 4445035
ChEBI CHEBI:52476
SMILES [Na+].[O-]C1=CC=CC=C1

Specifications

Melting Point >300°C
Odor Phenol-like
Linear Formula C6H5ONa
Quantity 250 g
UN Number UN2905
Sensitivity Air Sensitive
Solubility Information Very soluble in water,soluble in alcoholSoluble in water,acetone and alcohol.
IUPAC Name sodium benzenolate
Formula Weight 116.09
Percent Purity 98%
Chemical Name or Material Sodium phenoxide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: PHENOLATES, SOLID
EINECSNumber 205-347-3
RTECSNumber SM8780000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.