Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phenylpyruvate, 98%

Catalog No. p-7037921
Click to view available options
Quantity:
5 g
25 g

CAS: 114-76-1 | C9H7NaO3 | 186.142 g/mol

Phenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.

Solubility
Soluble in water

Notes
Store at 2-8°C. Avoin oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114-76-1
Molecular Formula C9H7NaO3
Molecular Weight (g/mol) 186.142
MDL Number MFCD00002590
InChI Key MQGYVGKMCRDEAF-UHFFFAOYSA-M
Synonym sodium phenylpyruvate, phenylpyruvic acid sodium salt, sodium 2-oxo-3-phenylpropanoate, unii-7528cw33ww, sodium ?-phenylpyruvate, phenylpyruvic acid, sodium salt, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, 2-oxo-3-phenylpropionic acid, 3-phenyl-2-oxopropanoic acid, sodium salt
PubChem CID 23667645
IUPAC Name sodium;2-oxo-3-phenylpropanoate
SMILES C1=CC=C(C=C1)CC(=O)C(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 4770978
Solubility Information Soluble in water
Formula Weight 186.14
Percent Purity 98%
Chemical Name or Material Sodium phenylpyruvate
EINECSNumber 204-053-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.