Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Phosphate, 0.2 M Buffer Solution, pH 7.0, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
Description
Applications
Sodium phosphate is widely used as a reagent with high buffering capacity for molecular biology, biochemistry, and chromatography
Solubility
Fully miscible.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Chemical Name or Material | Sodium phosphate buffer solution |
Concentration | 0.2 M |
pH | 7.0 |
Physical Form | Liquid |
Quantity | 1 L |
MDL Number | MFCD00146206 |
Solubility Information | Fully miscible. |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction