Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Phosphate, 0.2 M Buffer Solution, pH 7.2, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
Description
Applications
Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
Solubility
Fully miscible.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Chemical Name or Material | Sodium phosphate buffer solution |
Concentration | 0.2 M |
pH | 7.2 |
Physical Form | Liquid |
Quantity | 1 L |
MDL Number | MFCD00146206 |
Solubility Information | Fully miscible. |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction