Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Phosphate, 0.2 M Buffer Solution, pH 7.4, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
Description
Applications
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately. It is also used in the preparation of mobile phase in high performance liquid chromatography (HPLC).
Solubility
Miscible with water. Immiscible with alcohol.
Notes
Incompatible with strong acids.

Specifications
Specifications
Chemical Name or Material | Sodium phosphate buffer solution |
Concentration | 0.2 M |
pH | 7.4 |
Physical Form | Liquid |
Quantity | 1 L |
MDL Number | MFCD00146206 |
Solubility Information | Miscible with water. Immiscible with alcohol. |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction