Learn More
Sodium Phosphate, 0.2 M Buffer Solution, pH 8.0, PROMO, Thermo Scientific Chemicals
Description
Applications
Phosphate buffer is a multi-purpose buffer used for many molecular biology procedures. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.
Solubility
Fully miscible in water.
Notes
Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Chemical Name or Material | Sodium phosphate buffer solution |
Concentration | 0.2 M |
pH | 8.0 |
Physical Form | Liquid |
Quantity | 500 mL |
MDL Number | MFCD00146206 |
Solubility Information | Fully miscible in water. |
RUO - Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.