Learn More
Sodium Phosphate, 0.2 M Buffer Solution, pH 8.5, PROMO, Thermo Scientific Chemicals
Description
Applications
Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.
Solubility
Soluble in water (121 mg/mL at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong acids.

Specifications
Specifications
Chemical Name or Material | Sodium phosphate buffer solution |
Concentration | 0.2 M |
pH | 8.5 |
Physical Form | Liquid |
Quantity | 1 L |
MDL Number | MFCD00146206 |
Solubility Information | Soluble in water. |
RUO - Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.