Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Phosphate, 0.5 M Buffer Solution, pH 8.5, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
500 mL
Specifications
Specifications
Chemical Name or Material | Sodium phosphate buffer solution |
Concentration | 0.5 M |
pH | 8.5 |
Physical Form | Liquid |
Quantity | 500 mL |
MDL Number | MFCD00146206 |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction