Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate, Dibasic, 12-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

10039-32-4
H25Na2O16P
358.137
DGLRDKLJZLEJCY-UHFFFAOYSA-L
61456

High quality chemicals for laboratory and specialized industrial use

SMILES O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
IUPAC Name disodium;hydrogen phosphate;dodecahydrate
Formula Weight 358.14
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.50kg
Quantity 500g
Chemical Name or Material Sodium Phosphate, Dibasic, 12-Hydrate
Grade BAKER ANALYZED™ Reagent