Learn More
Sodium Phosphate Dibasic, ≥99.999% (metals basis), Honeywell™ Fluka™ TraceSELECT™
Shop All Honeywell Research Chemicals ProductsDescription
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Specifications
Specifications
| Quantity | 100 g |
| Linear Formula | Na2HPO4 |
| UN Number | UN3077 |
| Formula Weight | 141.96g/mol |
| Percent Purity | ≥99.999% (metals basis) |
| Packaging | Poly bottle |
| Chemical Name or Material | Sodium phosphate dibasic |
Safety and Handling
| DOTInformation | 9/PGIII |
| RTECSNumber | 231-448-7 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.