Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Dibasic Anhydrous, Purified (Disodium Hydrogen Phosphate), Reagents

Catalog No. 01336987
Click to view available options
Quantity:
500 g
2.5 kg
10 kg

Sodium Phosphate Dibasic Anhydrous, Purified (Disodium Hydrogen Phosphate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym Disodium Hydrogen Phosphate
IUPAC Name disodium hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 10 kg
Grade Reagent
Packaging Poly Pail
Specific Gravity 2.066
Chemical Name or Material Sodium Phosphate Dibasic Anhydrous, Purified (Disodium Hydrogen Phosphate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.