Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phosphate, dibasic dodecahydrate, 99%, extra pure, Acros Organics

Click to view available options
Quantity:
1kg
5kg
Packaging:
Plastic bottle
10039-32-4
HNa2O412H2O
358.13
DGLRDKLJZLEJCY-UHFFFAOYSA-L
disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12
61456
disodium;hydrogen phosphate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
10039-32-4
HNa2O412H2O
358.13
DGLRDKLJZLEJCY-UHFFFAOYSA-L
disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12
61456
disodium;hydrogen phosphate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
Assay Percent Range 99%
Linear Formula Na2HPO4·12H2O
Merck Index 15,8789
Solubility Information Solubility in water: 218g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 358.13
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Solubility Solubility in water: 218g/L (20°C). Other solubilities: practically insoluble in alcohol
Melting Point 35.0°C
Quantity 1kg
Chemical Name or Material Sodium phosphate, dibasic dodecahydrate
Show More Show Less