Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Phosphate Dibasic Heptahydrate (Colorless-to-White Crystals), Fisher BioReagents™
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Description
Sodium Phosphate Dibasic Heptahydrate (Colorless-to-White Crystals)
This buffer component is commonly used in tissue culture and molecular biology applications.

Specifications
Specifications
Melting Point | 48°C |
Color | White |
pH | 9.0 to 9.3 |
Physical Form | Solid |
Quantity | 500 g |
Assay Percent Range | 98.0 to 102.0 % |
Merck Index | 15, 8789 |
Formula Weight | 268.07 |
Percent Purity | 98.0 to 102.0% |
Identification | Pass Test |
Show More |
Safety and Handling

Recommended Storage | RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction